Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein, 0.2% w/v in Methanol, Reagents

Catalog No. 01333446
Click to view available options
Quantity:
1 L

Thymolphthalein, 0.2% w/v in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1, 125-20-2
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym 5',5''-Diisopropyl-2',2''-dimethylphenolphthalein
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 65°C
Percent Purity 0.2%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Color Colorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Thymolphthalein, 0.2% w/v in Methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.