Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein, ACS Reagent, Reagents

Catalog No. 01336930
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-930 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-930 Supplier Reagents Holdings Llc Supplier No. C233845010C1
Only null left
Add to Cart
Add to Cart

Thymolphthalein, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.54
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
Synonym 5',5''-Diisopropyl-2',2''-dimethylphenolphthalein
IUPAC Name 3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
SMILES CC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C

Specifications

Melting Point 245°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 0.92
pH Color Changes Colorless (pH <8.8) to Pale, grayish-blue (pH 9.4) to Blue (pH 9.9) to Intense Blue (pH >10.5)
Quantity 10 g
Physical Form Solid
Chemical Name or Material Thymolphthalein, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.