Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%

Catalog No. AC427670010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC427670010

Supplier: Thermo Scientific Chemicals 427670010

Only null left
Add to Cart

CAS: 129946-88-9 | C14H8F6O3S2 | 402.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129946-88-9
Molecular Formula C14H8F6O3S2
Molecular Weight (g/mol) 402.33
MDL Number MFCD00236132
InChI Key QXXHXTRTGZBOGD-UHFFFAOYSA-M
Synonym 5-trifluoromethyl-5h-dibenzo b,d thiophen-5-ium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, 5-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium triflate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulphonate, trifluoromethanesulfonate; 5-trifluoromethyl dibenzothiophen-5-ium, pubchem16425, acmc-20a6nt, c13h8f3s.chf3o3s, s-trifluoromethyl dibenzothiophenium trifluoromet
PubChem CID 2777507
IUPAC Name trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
SMILES [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 150°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 402.33
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.