Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THYN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558522 Supplier Invitrogen™ Supplier No. PA558522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFFYHSNCKE PGIAGLMKIV KEAYPDHTQF EKNNPHYDPS SKEDNPKWSM VDVQFVRMMK RFIPLAELKS YHQAHKATGG PL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THYN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THYN1
Gene Accession No. Q6P3E0, Q91YJ3, Q9P016
Gene Alias D730042P09Rik; HSPC144; MDS012; mThy28; My0054; MY105; Thy28; THY28KD; Thymocyte nuclear protein 1; thymocyte protein thy28; Thyn1
Gene Symbols THYN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THYN1. Recombinant protein control fragment (Product #RP-97415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29087, 300470, 77862
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.