Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Antibody (5E6cc), Alexa Fluor™ 647

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108083AF647
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
Thyroglobulin | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TG | |
Thyroglobulin from human thyroid gland. | |
0.2 mL | |
Cancer, Cell Biology | |
7038 | |
Human | |
Purified |
ELISA | |
5000000 | |
ELISA, Sandwich ELISA | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This does not interfere with anti-thyroglobulin autoantibodies. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction