Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Antibody (6E1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233096V
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Thyroglobulin | |
Monoclonal | |
DyLight 405 | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
Mouse | |
660 kDa | |
0.1 mL | |
Cancer, Cell Biology | |
7038 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
60 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
TG | |
Thyroglobulin isolated from human thyroid glands was used as the immunogen for the antibody. | |
Protein A or G purified | |
RUO | |
Primary | |
Thyroglobulin is a 660kDa dimeric pre-protein with multiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction