Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Antibody (rTGB/4744), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308264JF646
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
Thyroglobulin | |
Monoclonal | |
Janelia Fluor 646 | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
Human thyroid follicular cells | |
100 μg | |
Cancer, Cell Biology | |
7038 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
rTGB/4744 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction