Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Antibody (TGB/1970R), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308265FR
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Thyroglobulin | |
Monoclonal | |
DyLight 680 | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
Recombinant full-length human Thyroglobulin protein (Uniprot: P01266) | |
100 μg | |
Cancer, Cell Biology | |
7038 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
TGB/1970R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction