Learn More
Invitrogen™ Thyroglobulin Polyclonal Antibody

Description
Immunogen sequence: KMCSADYADL LQTFQVFILD ELTARGFCQI QVKTFGTLVS IPVCNNSSVQ VGCLTRERLG VNVTWKSRLE DIPVASLPDL HDIERALVGK DLLGRFTDLI QSGSFQLHLD SKTFPAETIR FLQGDHFGTS PRTWFGCSEG FYQVLTSEAS QDGLGCVKCP EGS Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 76%.
Specifications
Specifications
| Antigen | Thyroglobulin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TG |
| Gene Accession No. | P01266 |
| Gene Alias | AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor |
| Gene Symbols | TG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.