Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid Cancer Marker Monoclonal Antibody (373E1), Invitrogen™

Catalog No. PIMA512629
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512629 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512629 Supplier Thermo Scientific Supplier No. MA512629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone 373E1 is specific for the keratan sulfate bearing molecular complex. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10-20 min followed by cooling at room temperature for 20 min. Recommended positive control: papillary thyroid carcinoma.

Papillary carcinoma is by far the most frequent malignant tumor of the thyroid with a threefold prevelance in females. Studies have shown that 100% of malignant thyroid lesions were found to contain the keratan sulfate (KS) bearing molecules, while these were virtually absent from benign tissues and other thyroid tumors, with the exception of 21% of follicular carcinoma cases analyzed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroid Cancer Marker
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 373E1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 15mM sodium azide
Host Species Mouse
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.