Learn More
Thyroid Hormone Receptor beta 1 Polyclonal Antibody, Rockland™

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with human THRB protein. A BLAST analysis was used to suggest cross-reactivity with THRB from mouse, human and rat based on a 100% homology with the immunizing sequence. Cross-reactivity with THRB from other sources has not been determined.
Specifications
Specifications
| Antigen | Thyroid Hormone Receptor beta 1 |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 |
| Gene | THRB |
| Gene Accession No. | P10828, P18113, P37242 |
| Gene Alias | c-erbA-2; c-erbAbeta; c-erbA-beta; ERBA 2; ERBA BETA; Erba2; ERBA-BETA; GRTH; LOW QUALITY PROTEIN: thyroid hormone receptor beta; MGC126109; MGC126110; NR1A2; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; PRTH; RATT3REC; T3R[b]; T3Rbeta; T3rec; THR1; Thrb; Thrb1; Thrb2; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor beta 3; thyroid hormone receptor beta2delta; thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2); thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR; TR beta; TR beta 1; TR-B1C-ERBA-2; TRbeta; TRbeta2Delta |
| Gene Symbols | THRB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.