Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroid Peroxidase Antibody (313), DyLight 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290193UV
Description
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for ELISA.Specifications
| Thyroid Peroxidase | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 313 | |
| ELISA | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Thyroid Peroxidase (P07202-1; Met1-Arg846,257Ser/Ala,725Pro/Thr). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 7173 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction