Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroid Peroxidase Antibody (TPO/6417R), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308762AF532
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Thyroid Peroxidase | |
Monoclonal | |
Alexa Fluor 532 | |
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
Recombinant fragment of human Thyroid Peroxidase (around aa 685-804) (Exact sequence is proprietary) (Uniprot: P07202 ) | |
100 μg | |
Lipid and Metabolism | |
7173 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
TPO/6417R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction