Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroid Peroxidase Antibody (TPO/7088R) [Janelia Fluor™ 525], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321049JF525
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Thyroid Peroxidase | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Recombinant fragment of human Thyroid Peroxidase (around aa 600-900) (Exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 7173 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TPO/7088R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction