Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thyroid Peroxidase Monoclonal Antibody (08)
GREENER_CHOICE

Catalog No. PIMA529602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529602 Supplier Invitrogen™ Supplier No. MA529602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Thyroid peroxidase/TPO.

Thyroid Peroxidase is a gene that encodes for a membrane-bound glycoprotein, which functions as an enzyme and plays a crucial role in the proper functioning of the thyroid gland. This protein is responsible for the iodination of tyrosine residues in thyroglobulin and the formation of phenoxy-ester between pairs of iodinated tyrosines, which leads to the production of thyroid hormones such as thyroxine and triiodothyronine. Mutations in this gene are known to cause several disorders related to thyroid hormone production, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Several transcript variants encoding distinct isoforms have been identified for this gene, but the full-length nature of some variants remains undetermined. Diseases associated with TPO include Thyroid Dyshormonogenesis 2A and Congenital Hypothyroidism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroid Peroxidase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 08
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TPO
Gene Accession No. P07202
Gene Alias EC 1.11.1; LOW QUALITY PROTEIN: thyroid peroxidase; MSA; TDH2A; thyroid microsomal antigen; thyroid peroxidase; thyroid peroxidase (332 AA); thyroperoxidase; TPO; TPX; TPXEC 1.11.1.8
Gene Symbols TPO
Host Species Mouse
Immunogen Recombinant Human Thyroid peroxidase/TPO Protein (aa 1-846).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.