Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid peroxidase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980855
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0855 Supplier Bioss Supplier No. BS10406R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Thyroid Peroxidase is a transmembrane protein of 107 kDa containing a heme prosthetic group and it is present on the apical surface of the thyroid follicular cells. It is the primary enzyme involved in thyroid hormone synthesis. Malignant thyroid tumors exhibit an anomaly in Thyroid Peroxidase resulting in lower antibody affinity. This antibody may be useful aid in the differentiation between the benign and malignant thyroid tumors.

Specifications

Antigen Thyroid peroxidase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TPO
Gene Accession No. P07202
Gene Alias EC 1.11.1; LOW QUALITY PROTEIN: thyroid peroxidase; MSA; TDH2A; thyroid microsomal antigen; Thyroid peroxidase; thyroid peroxidase (332 AA); thyroperoxidase; TPO; TPX; TPXEC 1.11.1.8
Gene Symbols TPO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Thyroid peroxidase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7173
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.