Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid receptor-interacting protein 11 Antibody, Novus Biologicals™
SDP

Catalog No. NB393977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393977 25 μL
NBP257415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393977 Supplier Novus Biologicals Supplier No. NBP25741525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Thyroid receptor-interacting protein 11 Polyclonal specifically detects Thyroid receptor-interacting protein 11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Thyroid receptor-interacting protein 11
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias CEV14TR-interacting protein 11, Clonal evolution-related gene on chromosome 14 protein, GMAP-210ACG1A, Golgi-associated microtubule-binding protein 210, Golgi-microtubule-associated protein of 210 kDa, thyroid hormone receptor interactor 11, thyroid receptor-interacting protein 11, TRIP-11, Trip230
Gene Symbols TRIP11
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MLDDVQKKLMSLANSSEGKVDKVLMRNLFIGHFHTPKNQRHEVLRLMGSILGVRREEMEQLFHDDQGSVTRWMTGWLGGGSKSVPNTPLRPNQQSV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9321
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.