Learn More
Invitrogen™ Thyroxine Monoclonal Antibody (1H1)

Description
This antibody reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%.
Specifications
Specifications
| Antigen | Thyroxine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1H1 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Mouse |
| Immunogen | Conjugate of purified thyroxine with BSA. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.