Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thyroxine Monoclonal Antibody (XM212)
GREENER_CHOICE

Catalog No. PIMA122093
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA122093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA122093 Supplier Invitrogen™ Supplier No. MA122093
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%.

Thyroxine is a hormone produced by the thyroid glands to regulate metabolism by controlling the rate of oxidation in cells. Cases of hypothyroidism, where the gland is insufficiently active, can be treated by administration of thyroxine or a combination of thyroxine and triiodothyronine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroxine
Applications ELISA
Classification Monoclonal
Clone XM212
Concentration 6.0 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide; pH 7.5
Host Species Mouse
Immunogen Conjugate of purified thyroxine with BSA.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical, Mammalia
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.