Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIA-1 Monoclonal Antibody (OTI1C8)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526475
Description
TIA-1 Monoclonal Antibody for Western Blot, IHC (P)
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
Specifications
| TIA-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31483 | |
| TIA1 | |
| Full length human recombinant protein of TIA1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C8 | |
| Unconjugated | |
| TIA1 | |
| 2310050N03Rik; AI256674; cytotoxic granule-associated RNA binding protein 1; cytotoxic granule-associated RNA-binding protein 1; mTIA-1; Nucleolysin TIA-1; nucleolysin TIA-1 isoform p40; p40 TIA 1; p40-TIA-1; p40-TIA-1 (containing p15-TIA-1); RNA-binding protein TIA-1; T-cell-restricted intracellular antigen-1; Tia; TIA 1; TIA1; TIA-1; TIA1 cytotoxic granule-associated RNA binding protein; TIA1 protein; TIAL1; TIAR; WDM | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7072 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction