Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIA1 Antibody (TIA1/1313), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254406MFV450
Description
TIA1 Monoclonal antibody specifically detects TIA1 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
TIA1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
TIA1/1313 | |
Flow Cytometry, Flow (Intracellular) | |
cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein | |
Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380) (Uniprot: P31483) | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Immunology | |
7072 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction