Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIA1 Mouse anti-Human, Clone: OTI1C8, lyophilized, TrueMAB™
SDP

Catalog No. 501665238
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-5238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-5238 Supplier OriGene Supplier No. CF800659
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs , and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.

Specifications

Antigen TIA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C8
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TIA1
Gene Accession No. P31483
Gene Alias TIA-1, WDM
Gene Symbols TIA1
Host Species Mouse
Immunogen Full length human recombit protein of human TIA1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7072
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.