Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100757 Supplier Invitrogen™ Supplier No. PA5100757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIAL1.

The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs (RRMs), and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIAL1
Gene Accession No. P70318, Q01085
Gene Alias 5330433G13Rik; aging-associated gene 7 protein; AL033329; mTIAR; Nucleolysin TIAR; RNA binding protein TIAR (TIA-1 related); TCBP; T-cluster binding protein; Tia1 cytotoxic granule-associated RNA binding protein-like 1; TIA-1-related nucleolysin; TIA-1-related protein; Tial 1 cytotoxic granule-associated RNA- binding protein-like 1; TIAL1; Tial1 cytotoxic granule-associated RNA binding protein-like 1; Tial1 cytotoxic granule-associated RNA-binding protein-like 1; TIAR
Gene Symbols TIAL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIAL1(Accession Q01085), corresponding to amino acid residues A254-Q304.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21843, 361655, 7073
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.