Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562325 Supplier Invitrogen™ Supplier No. PA562325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSPSSPFRE QSFLCAAGDA GEESRVQVLK NEVRRGSPVL LGWVEQAYAD KCV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 28%.

TIAF-1 (TGFB1-induced anti-apoptotic factor 1), also known as MYO18A, is a TGFB1-induced anti-apoptotic factor that functions to inhibit the cytotoxic effects of TNF-alpha, TRADD and FADD. It localizes to the nucleus and significantly up-regulated in activated helper T lymphocytes (TH2) in chronic kidney and liver allograft rejection. Suppression of TIAF1 expression by siRNA prevents UV irradiation-mediated p53 phosphorylation and nuclear translocation. TIAF1 is essential in the p53- and WOX1-mediated cell death and shows an aggregation-dependent control of tumor progression and metastasis, and regulation of cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIAF1
Gene Accession No. O95411
Gene Alias 12 kDa TGF-beta-1-induced antiapoptotic factor; MAJN; molecule associated with Jak-3 N-terminal; MYO18A; MYSPDZ; SPR210; TGFB1-induced anti-apoptotic factor 1; TGF-beta-1-induced antiapoptotic factor 1; TIAF; TIAF1
Gene Symbols TIAF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIAF1. Recombinant protein control fragment (Product #RP-100356).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.