Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiagabine hydrochloride, 98+%, Thermo Scientific™

Catalog No. AC456200010
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 145821-59-6
Molecular Formula C20H26ClNO2S2
Molecular Weight (g/mol) 412.00
MDL Number MFCD07369025
InChI Key YUKARLAABCGMCN-PKLMIRHRSA-N
Synonym tiagabine hydrochloride, tiagabine hcl, gabitril, abbott 70569.hcl, unii-dqh6t6d8oy, dqh6t6d8oy, abbott-70569.hcl, r-1-4,4-bis 3-methylthiophen-2-yl but-3-en-1-yl piperidine-3-carboxylic acid hydrochloride, --r-1-4,4-bis 3-methyl-2-thienyl-3-butenyl nipecotic acid, hydrochloride
PubChem CID 91274
ChEBI CHEBI:85388
IUPAC Name (3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-enyl]piperidine-3-carboxylic acid;hydrochloride
SMILES [H+].[Cl-].CC1=C(SC=C1)C(=CCCN1CCC[C@H](C1)C(O)=O)C1=C(C)C=CS1

Specifications

Melting Point 192.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 412.02
Percent Purity 98+%
Chemical Name or Material Tiagabine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.