Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIAL1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204047JF646
Description
TIAL1 Polyclonal antibody specifically detects TIAL1 in Human samples. It is validated for Western BlotSpecifications
TIAL1 | |
Polyclonal | |
Western Blot | |
aging-associated gene 7 protein, MGC33401, TCBP, T-cluster binding protein, TIA1 cytotoxic granule-associated RNA binding protein-like 1, TIA1 cytotoxic granule-associated RNA-binding protein-like 1, TIA-1 related protein, TIA-1-related nucleolysin, TIA-1-related protein, TIARnucleolysin TIAR | |
The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human TIA-1-Related Protein using the numbering given in entry NP_003243.1 (GeneID 7073). | |
0.1 mL | |
Apoptosis | |
7073 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction