Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIAL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596501
Description
Immunogen sequence: EGHVVKCYWG KESPDMTKNF QQVDYSQWGQ WSQVYGNPQQ YGQYMANGWQ VPPYGVYGQP WNQQGFGVDQ SPSAAWMGGF GAQPPQGQAP PPVIPPPNQA GYGMASYQTQ; Positive Samples: A-549, HeLa, K-562; Cellular Location: Cytoplasm, Cytoplasmic granule, Nucleus.
The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs (RRMs), and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.
Specifications
TIAL1 | |
Polyclonal | |
Unconjugated | |
TIAL1 | |
5330433G13Rik; aging-associated gene 7 protein; AL033329; mTIAR; Nucleolysin TIAR; RNA binding protein TIAR (TIA-1 related); TCBP; T-cluster binding protein; Tia1 cytotoxic granule-associated RNA binding protein-like 1; TIA-1-related nucleolysin; TIA-1-related protein; Tial 1 cytotoxic granule-associated RNA- binding protein-like 1; TIAL1; Tial1 cytotoxic granule-associated RNA binding protein-like 1; Tial1 cytotoxic granule-associated RNA-binding protein-like 1; TIAR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21843, 361655, 7073 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70318, Q01085 | |
TIAL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 266-375 of human TIAL1 (NP_0032431). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction