Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TIAM-1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1488MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1488MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1488MI Supplier MilliporeSigma™ Supplier No. ABN1488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

T-lymphoma invasion and metastasis-inducing protein 1 (UniProt Q13009; also known as TIAM-1) is encoded by the TIAM1 gene (Gene ID 7074) in human. TIAM-1 is a ubiquitously expressed guanine nucleotide exchange factor (GEF) that specifically activates the Rho-like GTPase Rac1 in response to growth factors and cell-substrate interaction. TIAM-1 associates with GTP-bound Ras and functions as an effector that mediates Rac activation by Ras. TIAM-1 promotes the formation of adherens junctions and the induction of an epithelial-like phenotype in mesenchymal cells, and is essential for the formation and maintenance of cadherin-based adhesions. Tiam1-knockdown in mice protects against Ras-induced skin carcinogenesis as a result of increased apoptosis and reduced proliferation/growth of keratinocytes. However, the fewer skin tumors developed progress more frequently to malignancy in Tiam1-knockout mice, indicating that loss of Tiam1 stimulates malignant conversion. Cellular TIAM-1 is targeted for proteasome-dependent degradation by the SCFbTrCP ubiquitin ligase in cooperation with casein kinase 1 and the MEK/ERK pathway upon mitogenic stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAM-1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene TIAM1
Gene Accession No. Q13009
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human TIAM-1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_003244
Test Specificity This rabbit polyclonal antibody targets a C-terminal region of human and mouse TIAM-1 sequences reported by UniProt (Q13009 and Q60610).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.