Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIAM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002301AF750
Description
TIAM1 Polyclonal antibody specifically detects TIAM1 in Human, Mouse samples. It is validated for Western BlotSpecifications
TIAM1 | |
Polyclonal | |
Western Blot | |
FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1 | |
The immunogen recognized by this antibody maps to a region between residues 1550 and the C-terminus (residue 1591) of human T-cell lymphoma invasion and metastasis 1 using the numbering given in entry NP_003244.1 (GeneID 7074) | |
0.1 mL | |
Cancer | |
7074 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction