Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIAM1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP275096
Description
TIAM1 Polyclonal specifically detects TIAM1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
TIAM1 | |
Polyclonal | |
Unconjugated | |
TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide | |
TIAM1 | |
Peptide with sequence C-EDFAPSRKLNTEI, from the C Terminus of the protein sequence according to NP_003244; NP_001340613.1; NP_001340614.1; NP_001340615.1; NP_001340616.1. | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/ml | |
Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Peptide ELISA | |
FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1 | |
Goat | |
Antigen Affinity-purified | |
Cancer | |
7074.0 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction