Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580130 Supplier Invitrogen™ Supplier No. PA580130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 whole cell.

Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP-dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Activates RAC1, CDC42, and to a lesser extent RHOA. Required for normal cell adhesion and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAM1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene TIAM1
Gene Accession No. Q13009
Gene Alias AI847750; D16Ium10; D16Ium10e; human T-lymphoma invasion and metastasis inducing TIAM1 protein; T cell lymphoma invasion and metastasis 1; T-cell lymphoma invasion and metastasis 1; Tiam1; TIAM-1; T-lymphoma invasion and metastasis-inducing protein 1
Gene Symbols TIAM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human TIAM1 (1518-1554aa ASVDRDLQERLQATSISQRERGRKTLDSHASRMAQLK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7074
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.