Learn More
TIAM1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300099A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP-dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Activates RAC1, CDC42, and to a lesser extent RHOA. Required for normal cell adhesion and cell migration.Specifications
| TIAM1 | |
| Polyclonal | |
| Unconjugated | |
| TIAM1 | |
| TIAM-1, TIAM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21844, 7074 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q13009, Q60610 | |
| Tiam1 | |
| Between 1550 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.