Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142639
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142639 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142639 Supplier Invitrogen™ Supplier No. PA5142639
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform a (NP_036586.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP- dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Mediates extracellular laminin signals to activate Rac1, contributing to neurite growth. Involved in lamellipodial formation and advancement of the growth cone of embryonic hippocampal neurons. Promotes migration of neurons in the cerebral cortex. When overexpressed, induces membrane ruffling accompanied by the accumulation of actin filaments along the altered plasma membrane. Activates specifically RAC1, but not CDC42 and RHOA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Tiam2
Gene Accession No. Q8IVF5
Gene Alias 3000002F19Rik; KIAA2016; mKIAA2016; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; SIF and TIAM1-like exchange factor; Stef; T cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; TIAM2; TIAM-2; T-lymphoma invasion and metastasis-inducing protein 2
Gene Symbols Tiam2
Host Species Goat
Immunogen Peptide with sequence TLQGSKNHSNT-C (aa 9-19).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26230
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.