Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Environstar MP Coveralls

Catalog No. 19130201D Shop All Tians International Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Neck Style:
Collared
Hooded
Waist Style:
Elastic
Non-elastic
Ankle Style:
Elastic
This item is not returnable. View return policy
This item is not returnable. View return policy

Protective apparel manufactured in climate-controlled facility

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Certifications/Compliance ANSI ISEA 101-1996
For Use With (Application) Protects against petroleum Oils, pesticides, Bleach and Sodium Hydroxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.