Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tians epic™ High Performance Static Dissipative Cleanroom Frocks
Shop All Tians International Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Lint-free film coated frock
- Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
- Features include a snap front, elastic wrists
- Mandarin snap collar, no pockets
- Fabric is constructed from a blend of polyethylene and polypropylene
- Finished fabric contains a proprietary additive making it static dissipative
- For use in ISO 6 Cleanroom Class environments
- Neatly folded and cleanroom packed
- 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case
Specifications
Specifications
| Product Type | Controlled Environments Frock |
| Size | X-Large |
| Material | Polypropylene, Polyethylene |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Collar Type | Mandarin |
| Size (English) Chest | 11 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction