Tians™ Epic™ SafeTrack-HD™ High Traction Boot Covers

Suitable for cleanroom, critical and industrial environments

Manufacturer: Tians International 534742L

 View more versions of this product

Catalog No. 19-138-921

  Check Availability
Add to Cart

    Specifications

    Size Large
    Sole Type Non-skid
    Product Type High Traction Shoe Covers
    Material Polyethylene, Polypropylene
    Disposable Yes
    For Use With (Application) Ideal for many cleanroom, critical and industrial environment
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit