Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Epic™ Heavy Polyethylene Shoe Covers

Catalog No. 191600526 Shop All Tians International Products
Click to view available options
Size:
Large
3X-Large
This item is not returnable. View return policy

Catalog No. 19-160-0526

Supplier: Tians 747773L

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Shoe covers constructed from a heavyweight polyethylene film for added strength and durability.

  • Lint-free, latex-free
  • Internally sewn elastic bottom seam
  • Heavyweight polyethylene film
  • Embossed texture for added skid resistance
  • For use in critical and low particulate clean environments and many general industrial applications
  • Fabric is suitable for exposure to bloodborne pathogens and some hazardous liquids
  • 6 mil material thickness
  • Suitable for ISO Cleanroom Class 5

Specifications

Size Large
Material Polyethylene
Latex Free Yes
Color Blue
Seam Type Serged
Thickness 6 mil
For Use With (Application) Critical and low particulate clean environments and many general industrial applications
Packaging Quantity 100 pieces per sealed inner polybag, 3 polybags per sealed master bag, 300 pieces per case
Product Type Controlled Environments Shoe Covers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.