Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIAR Mouse anti-Human,Monkey,Mouse, Unconjugated, Clone: 6E3, MilliporeSigma™

Catalog No. MABF26512MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF26512MI 25 μg
MABF26511MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF26512MI Supplier MilliporeSigma Supplier No. MABF265125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIAR Monoclonal antibody specifically detects TIAR in Human,Monkey,Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAR
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6E3
Conjugate Unconjugated
Gene Accession No. NP_001029097.1
Gene Alias TIA-1 related protein;Nucleolysin TIAR
Gene Symbols TIAL1
Host Species Mouse
Immunogen GST-tagged Full-length recombinant human Nucleolysin TIAR protein.
Molecular Weight of Antigen ∼42 kDa observed; 41.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 6E3 is a mouse monoclonal antibody that specifically detects TIAR protein.
Target Species Human, Monkey, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.