Learn More
TIAR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303613A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs , and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.Specifications
TIAR | |
Polyclonal | |
Unconjugated | |
TIAL1 | |
TCBP, TIAR, T-cluster binding protein, TIA-1-related nucleolysin, aging-associated gene 7 protein, nucleolysin TIAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7073 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q01085 | |
TIAL1 | |
Between 325 and 375 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.