Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TICAM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523396
Description
TICAM2 Polyclonal Antibody for Western Blot, IHC (P)
TRAM, or TRIF-related adaptor molecule (TICAM2), plays an essential role in the MyD88-independent signaling of TLR4. The functional absence of TRAM in vascular endothelial cells restricts TLR4 signaling to MyD88-dependent pathway. Overexpression of TRAM cDNA in endothelial cells reconstituted LPS-induced Trif-dependent NF-kB activation and cxcl10 promoter activity.
Specifications
| TICAM2 | |
| Polyclonal | |
| Unconjugated | |
| TICAM2 | |
| B430113A10; cytoplasmic adaptor; MyD88-4; NF-kappa-B-activating protein 502; putative NF-kappa-B-activating protein 502; TAG; TICAM2; TICAM-2; TIR domain-containing adapter molecule 2; TIR domain-containing adapter protein; TIRAP3; Tirp; toll like receptor adaptor molecule 2; toll/interleukin-1 receptor (TIR) domain-containing adapter protein; toll/interleukin-1 receptor domain-containing protein; Toll-interleukin I receptor domain (TIR)-containing adaptor molecule (TICAM) 2; toll-like receptor adaptor molecule 2; toll-like receptor adaptor protein 3; Tram; Trif; TRIF-related adapter molecule; TRIF-related adapter molecule TRAM; TRIF-related adaptor molecule | |
| Rabbit | |
| Protein A | |
| RUO | |
| 225471, 353376, 364867, 539350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q2LGB7, Q86XR7, Q8BJQ4 | |
| TICAM2 | |
| A portion of amino acids 150-200 of TAG/TIRAP3b. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Marsupial | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction