Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TID1 Monoclonal Antibody (RS13), Invitrogen™

Catalog No. PIMA512267
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512267 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512267 Supplier Thermo Scientific Supplier No. MA512267

Mouse Monoclonal Antibody

MA5-12267 targets TID-1 in IP, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-12267 immunogen is recombinant hTid-1 protein.

TID1 is a human homolog of the Drosophila tumor suppressor lethal (2) tumorous imaginal discs, l(2)tid and encodes two mitochondrial matrix localized splice variants of human Tid-1 designated hTid-1S and hTid-1L. These proteins are the conserved members of the DnaJ family of proteins which act as cochaperones for mitochondrial Hsp70. They contain a conserved tetrahedrical J domain which binds to Hsp70 chaperones and activates their ATPase activity. Expression of hTid-1L increases apoptosis induced by DNA damaging agents as mitomycin-C and TNF-a. A J-domain mutant of hTid-1L can dominantly suppress apoptosis and in sharp contrast the J-domain mutant of hTid-1S increases apoptosis. Expression of hTid-1S and hTid-1L affects cytochrome c release from the mitochondria and caspase 3 activation, while activation of caspase 8 is unaffected. It is strongly suggested that these two splice variants exert their anti- and pro- apoptotic effects through discrete substrates and activities. Hence the relative abundance of these proteins or their substrates may allow the mitochondria to dampen or enhance the apoptotic signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TID1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RS13
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene DNAJA3
Gene Accession No. Q96EY1, Q99M87
Gene Alias 1200003J13Rik; 1810053A11Rik; C81173; DnaJ (Hsp40) homolog, subfamily A, member 3; DnaJ heat shock protein family (Hsp40) member A3; dnaJ homolog subfamily A member 3, mitochondrial; DnaJ protein Tid-1; Dnaja3; HCA57; Hepatocellular carcinoma-associated antigen 57; hTID-1; mTid-1; Tid1; Tid-1; Tid1l; Tumorous imaginal discs protein Tid56 homolog
Gene Symbols DNAJA3
Host Species Mouse
Immunogen Recombinant hTid-1 protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360481, 83945, 9093
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.