Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TID1 Polyclonal Antibody
TID1 Polyclonal Antibody
Supplier: Thermo Scientific PA598660
Description
TID1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

Specifications
TID1 | |
Polyclonal | |
Unconjugated | |
DNAJA3 | |
1200003J13Rik; 1810053A11Rik; C81173; DnaJ (Hsp40) homolog, subfamily A, member 3; DnaJ heat shock protein family (Hsp40) member A3; dnaJ homolog subfamily A member 3, mitochondrial; DnaJ protein Tid-1; Dnaja3; HCA57; Hepatocellular carcinoma-associated antigen 57; hTID-1; mTid-1; Tid1; Tid-1; Tid1l; Tumorous imaginal discs protein Tid56 homolog | |
Rabbit | |
Protein G | |
RUO | |
9093 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
3.64 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96EY1 | |
DNAJA3 | |
Recombinant Human DnaJ homolog subfamily A member 3, mitochondrial protein (86-184AA ). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction