Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-1 Antibody (6F12), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118610APCCY7
Description
Tie-1 Monoclonal specifically detects Tie-1 in Human samples. It is validated for Flow Cytometry.Specifications
Tie-1 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
TIE1 | |
Recombinant human soluble extracellular TIE-1 | |
0.1 mL | |
Primary | |
The monoclonal antibody will detect native human TIE-1 in ELISA experiments and on the surface of different human cell types. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
6F12 | |
Flow Cytometry | |
EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Protein Kinase | |
7075.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction