Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-1 Antibody (8D12B10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237270
Description
Tie-1 Monoclonal specifically detects Tie-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Tie-1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
Mouse | |
125 kDa | |
0.1 mL | |
Angiogenesis, Cancer, Protein Kinase | |
7075 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
8D12B10 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
TIE1 | |
Purified recombinant fragment of human TIE1 (AA: 385-607) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction