Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-2 Antibody (MM0018-21G7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | Tie-2 |
---|---|
Clone | MM0018-21G7 |
Concentration | LYOPH |
Dilution | Western Blot 1:500 - 1:1000, ELISA 1:200 - 1:400, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 |
Applications | Western Blot, Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017984
![]() |
Novus Biologicals
NB110609860.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11060986
![]() |
Novus Biologicals
NB11060986 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Tie-2 | |
LYOPH | |
Western Blot, Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Protein Kinase | |
Q02763 | |
7010 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified human extracellular domain of Tie2 fragment). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0018-21G7 | |
Western Blot 1:500 - 1:1000, ELISA 1:200 - 1:400, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
TEK | |
IgG1 | |
Protein G purified | |
Recognizes human Tie2, no cross-reactivity with human Tie1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title