Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-2 Antibody (tek9), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118614PEATT594
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry.Specifications
Tie-2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TEK | |
Recombinant human soluble extracellular TIE-2 | |
0.1 mL | |
Primary | |
The monoclonal antibody will detect native human TIE-2/tek in ELISA experiments and on the surface of different human cell types. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
tek9 | |
Flow Cytometry | |
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Phospho Specific, Protein Kinase | |
7010.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction