Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-2 Mouse anti-Human, Alexa Fluor 488, Clone: 7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314641AF488
This item is not returnable.
View return policy
Description
Tie-2 Monoclonal antibody specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
Tie-2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
7 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.2 mL | |
Angiogenesis, Cancer, Protein Kinase | |
7010 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction