Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-2 Rabbit anti-Human, Clone: 116, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00
Specifications
Antigen | Tie-2 |
---|---|
Clone | 116 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Sandwich ELISA Capture 1:250-1:2000 |
Applications | Flow Cytometry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Capture.Specifications
Tie-2 | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
Recombinant Human Tie-2 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
116 | |
Flow Cytometry, Immunofluorescence, ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Phospho Specific, Protein Kinase | |
0.2 um filtered solution in PBS | |
7010 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title