Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIE1 Monoclonal Antibody (4E7F11)
GREENER_CHOICE

Catalog No. PIMA530934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530934 Supplier Invitrogen™ Supplier No. MA530934
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Tie1.

Probable protein tyrosine-kinase transmembrane receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIE1
Applications ELISA
Classification Monoclonal
Clone 4E7F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TIE1
Gene Accession No. P35590
Gene Alias D430008P04Rik; JTK14; TIE; Tie1; Tie-1; tyrosine kinase receptor 1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin like and EGF like domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; tyrosine-protein kinase receptor Tie-1
Gene Symbols TIE1
Host Species Mouse
Immunogen Recombinant Human Tie1 Protein (Met1-Gln760).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.