Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585869
Description
TIE1 Polyclonal Antibody for Western Blot, ICC/IF
Probable protein tyrosine-kinase transmembrane receptor.
Specifications
TIE1 | |
Polyclonal | |
Unconjugated | |
TIE1 | |
D430008P04Rik; JTK14; TIE; Tie1; Tie-1; tyrosine kinase receptor 1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin like and EGF like domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; tyrosine-protein kinase receptor Tie-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21846, 7075, 89806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 25% Glycerol and 0.05% sodium azide; pH 7.4 | |
P35590, Q06806 | |
TIE1 | |
Synthetic peptide within Human TIE-1 aa 23-72. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction